Merck
  • Functional Expression of Gloeobacter Rhodopsin in Synechocystis sp. PCC6803.
Functional Expression of Gloeobacter Rhodopsin in Synechocystis sp. PCC6803.
Photochemistry and photobiology (2017-05-14)
Que Chen, Jos Arents, Srividya Ganapathy, Willem J de Grip, Klaas J Hellingwerf
ABSTRACT
Proteorhodopsins are light-driven proton pumps that occur widespread in Nature, where they function predominantly in environments with high incident irradiance. Their maximal absorbance is usually in the blue range, but can be extended into the (far)red range of the electromagnetic spectrum. Because they can be expressed heterologously, they may be exploited in studies aimed at increasing the efficiency of photosynthesis. Here we report further studies toward this goal, by comparing the expression of two different bacterial rhodopsins (Proteorhodopsin and Gloeobacter rhodopsin) in the model cyanobacterium Synechocystis sp. PCC6803. In particular, we investigated the pigments bound by the respective apo-opsins, and the oligomeric state of the corresponding holo-rhodopsins, both in Escherichia coli and in the cyanobacterial membranes. We conclude that the two proton-pumping rhodopsins are predominantly present in an oligomeric state (hexamers for Proteorhodopsin and trimers for Gloeobacter rhodopsin). Furthermore, Gloeobacter rhodopsin is able to bind an antenna carotenoid (in addition to retinal) and has the highest pumping rate at given light intensity. However, its lower expression level will decrease its physiological effectiveness. It remains to be established which of these two bacterial rhodopsins is best in stimulating the growth rate of its cyanobacterial host.
Sigma-Aldrich
Echinenone, ≥95.0% (HPLC)