- Systematic development of design of experiments (DoE) optimised self-microemulsifying drug delivery system of Zotepine.
Systematic development of design of experiments (DoE) optimised self-microemulsifying drug delivery system of Zotepine.
Journal of microencapsulation (2017-04-30)
Hitesh Dalvadi, Nikita Patel, Komal Parmar
PMID28452252
ABSTRACT
The aim of present investigation is to improve dissolution rate of poor soluble drug Zotepine by a self-microemulsifying drug delivery system (SMEDDS). Ternary phase diagram with oil (Oleic acid), surfactant (Tween 80) and co-surfactant (PEG 400) at apex were used to identify the efficient self-microemulsifying region. Box-Behnken design was implemented to study the influence of independent variables. Principal Component Analysis was used for scrutinising critical variables. The liquid SMEDDS were characterised for macroscopic evaluation, % Transmission, emulsification time and in vitro drug release studies. Optimised formulation OL1 was converted in to S-SMEDDS by using Aerosil