Merck
  • High-Performance Semicrystalline Poly(ether ketone)-Based Proton Exchange Membrane.
High-Performance Semicrystalline Poly(ether ketone)-Based Proton Exchange Membrane.
ACS applied materials & interfaces (2017-07-07)
Sinan Feng, Jinhui Pang, Xingwen Yu, Guibin Wang, Arumugam Manthiram
ABSTRACT
A novel semicrystalline poly(ether ketone) (PEK)-based proton exchange membrane (semi-SPEK-x) has been developed. Through a one-step sulfonation and hydrolysis, a poly(ether ketimine) precursor transforms into PEK and ion-conducting groups are introduced. With an ion-exchange capacity ranging from 1.49 to 2.00 mequiv g
Sigma-Aldrich
4,4′-Difluorobenzophenone, 99%
Sigma-Aldrich
Dichloroacetic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfolane, 99%
Sigma-Aldrich
Potassium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%