Merck
  • Copper(II) interaction with the Human Prion 103-112 fragment - Coordination and oxidation.
Copper(II) interaction with the Human Prion 103-112 fragment - Coordination and oxidation.
Journal of inorganic biochemistry (2017-03-07)
Gizella Csire, Lajos Nagy, Katalin Várnagy, Csilla Kállay
ABSTRACT
The prion protein (PrP) is a membrane-anchored cell surface glycoprotein containing 231 amino acids. It has been associated with a group of neurodegenerative disorders. Copper(II) interaction with the Human Prion 103-112 fragment and its mutants has been studied with various techniques. The studied human prion fragment contains both histidine and methionine residues, while methionine residues are systematically replaced or displaced in the studied mutants. pH-potentiometric, UV-Vis and circular dicroism spectroscopic techniques were applied to study the stoichiometry, stability and structure of the copper(II) complexes, while HPLC-MS and MS/MS were used for identifying the products of copper(II) catalyzed oxidation. The complex formation reactions of the studied ligands are rather similar; only 1:1 complexes are formed, where the imidazole nitrogen of the histidine residue is the main binding site beside the amide nitrogens of the peptide chain. The only difference is, that in the peptides which contain methionine in position 109, in addition to the (N
Sigma-Aldrich
Fmoc-Ser(tBu)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-His(Trt)-OH, ≥98.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
2,2′-(Ethylenedioxy)diethanethiol, 95%
Sigma-Aldrich
O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate, ≥97.0% (N)
Sigma-Aldrich
Fmoc-Lys(Boc)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Pro-OH, ≥90% (HPLC)
Sigma-Aldrich
Fmoc-Met-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Fmoc-Ala-OH, 95%