Merck
  • Interruption of beta-catenin signaling reduces neurogenesis in Alzheimer's disease.
Interruption of beta-catenin signaling reduces neurogenesis in Alzheimer's disease.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-05-22)
Ping He, Yong Shen
ABSTRACT
The neuronal loss associated with Alzheimer's disease (AD) affects areas of the brain that are vital to cognition. Although recent studies have shown that new neurons can be generated from progenitor cells in the neocortices of healthy adults, the neurogenic potential of the stem/progenitor cells of AD patients is not known. To answer this question, we compared the properties of glial progenitor cells (GPCs) from the cortices of healthy control (HC) and AD subjects. The GPCs from AD brain samples displayed reduced renewal capability and reduced neurogenesis compared with GPCs from HC brains. To investigate the mechanisms underlying this difference, we compared beta-catenin signaling proteins in GPCs from AD versus HC subjects and studied the effect of amyloid beta peptide (Abeta, a hallmark of AD pathology) on GPCs. Interestingly, GPCs from AD patients exhibited elevated levels of glycogen synthase kinase 3beta (GSK-3beta, an enzyme known to phosphorylate beta-catenin), accompanied by an increase in phosphorylated beta-catenin and a decrease in nonphosphorylated beta-catenin compared with HC counterparts. Furthermore. we found that Abeta treatment impaired the ability of GPCs from HC subjects to generate new neurons and caused changes in beta-catenin signaling proteins similar to those observed in GPCs from AD patients. Similar results were observed in GPCs isolated from AD transgenic mice. These results suggest that Abeta-induced interruption of beta-catenin signaling may contribute to the impairment of neurogenesis in AD progenitor cells.
Sigma-Aldrich
Anti-MAP2 Antibody, clone AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-Dlx2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Polysialic Acid-NCAM Antibody, clone 2-2B, ascites fluid, clone 2-2B, Chemicon®