Merck
  • PAX3-FOXO1A Expression in Rhabdomyosarcoma Is Driven by the Targetable Nuclear Receptor NR4A1.
PAX3-FOXO1A Expression in Rhabdomyosarcoma Is Driven by the Targetable Nuclear Receptor NR4A1.
Cancer research (2016-11-20)
Alexandra Lacey, Aline Rodrigues-Hoffman, Stephen Safe
ABSTRACT
Alveolar rhabdomyosarcoma (ARMS) is a devastating pediatric disease driven by expression of the oncogenic fusion gene PAX3-FOXO1A. In this study, we report overexpression of the nuclear receptor NR4A1 in rhabdomyosarcomas that is sufficient to drive high expression of PAX3-FOXO1A there. RNAi-mediated silencing of NR4A1 decreased expression of PAX3-FOXO1A and its downstream effector genes. Similarly, cell treatment with the NR4A1 small-molecule antagonists 1,1-bis(3-indolyl)-1-(p-hydroxy or p-carbomethoxyphenyl)methane (C-DIM) decreased PAX3-FOXO1A. Mechanistic investigations revealed a requirement for the NR4A1/Sp4 complex to bind GC-rich promoter regions to elevate transcription of the PAX3-FOXO1A gene. In parallel, NR4A1 also regulated expression of β1-integrin, which with PAX3-FOXO1A, contributed to tumor cell migration that was blocked by C-DIM/NR4A1 antagonists. Taken together, our results provide a preclinical rationale for the use of NR4A1 small-molecule antagonists to treat ARMS and other rhabdomyosarcomas driven by PAX3-FOXO1A. Cancer Res; 77(3); 732-41. ©2016 AACR.
Sigma-Aldrich
DIM-C-pPhCO2Me, ≥98% (HPLC)
Sigma-Aldrich
DIM-C-pPhOH, ≥98% (HPLC)