Merck
  • Diversity of the association of serum levels and genetic variants of MHC class I polypeptide-related chain A with liver fibrosis in chronic hepatitis C.
Diversity of the association of serum levels and genetic variants of MHC class I polypeptide-related chain A with liver fibrosis in chronic hepatitis C.
Oncotarget (2017-04-22)
Chung-Feng Huang, Ching-I Huang, Ming-Lun Yeh, Shu-Chi Wang, Kuan-Yu Chen, Yu-Min Ko, Ching-Chih Lin, Yi-Shan Tsai, Pei-Chien Tsai, Zu-Yau Lin, Shinn-Cherng Chen, Chia-Yen Dai, Jee-Fu Huang, Wan-Long Chuang, Ming-Lung Yu
ABSTRACT
Genetic variants of MHC class I polypeptide-related chain A (MICA) at rs2596542 have been associated with hepatocellular carcinoma. The linkage between serum MICA (sMICA) and liver fibrosis in chronic hepatitis C is elusive. Linear regression analysis revealed that sMICA were independently correlated to α-fetoprotein (β: 0.149; 95% confidence interval [CI]: 0.001, 0.003; P = 0.007)and MICA rs2596542 GG genotype (β: 0.209; 95% CI: 0.153, 0.483; P < 0.001). While patients were stratified by MICA genetic variants, advanced fibrosis was the only factor independently correlated to sMICA among A allele carriers (β: 0.234; 95% CI: 0.107, 0.543; P = 0.004) but not among non-A allele carriers. Logistic regression analysis revealed that factors associated with advanced liver fibrosis was sMICA (OR/CI: 2.996/1.428-6.287, P = 0.004) and platelet counts (OR/CI: 0.988/0.982-0.994, P < 0.001) in MICA rs2596542 A allele carriers. sMICA > 50 pg/mL provided a positive predictive value of 72 % in predicting advanced liver fibrosis (F3-4) and of 90% in significant fibrosis (> F2) in MICA rs2596542 A allele carriers. Serum level and single nucleotide polymorphism at rs2596542 of MICA were tested for the association with liver fibrosis in 319 biopsy proven chronic hepatitis C patients. Levels of sMICA were highly correlated to liver disease severity in chronic hepatitis C patients who carried the MICA rs738409 A allele. Patients possessing the genetic predisposition had a higher likelihood of progressed liver fibrosis if they expressed higher sMICA levels.