Merck
  • Enhanced generation of human induced pluripotent stem cells by ectopic expression of Connexin 45.
Enhanced generation of human induced pluripotent stem cells by ectopic expression of Connexin 45.
Scientific reports (2017-03-30)
Qiong Ke, Li Li, Xin Yao, Xingqiang Lai, Bing Cai, Hong Chen, Rui Chen, Zhichen Zhai, Lihua Huang, Kai Li, Anbin Hu, Frank Fuxiang Mao, Andy Peng Xiang, Liang Tao, Weiqiang Li
ABSTRACT
Somatic cells can be successfully reprogrammed into pluripotent stem cells by the ectopic expression of defined transcriptional factors. However, improved efficiency and better understanding the molecular mechanism underlying reprogramming are still required. In the present study, a scrape loading/dye transfer assay showed that human induced pluripotent stem cells (hiPSCs) contained functional gap junctions partially contributed by Connexin 45 (CX45). We then found CX45 was expressed in human embryonic stem cells (hESCs) and human dermal fibroblasts (hDFs) derived hiPSCs. Then we showed that CX45 was dramatically upregulated during the reprogramming process. Most importantly, the ectopic expression of CX45 significantly enhanced the reprogramming efficiency together with the Yamanaka factors (OCT4, SOX2, KLF4, cMYC - OSKM), whereas knockdown of endogenous CX45 expression significantly blocked cellular reprogramming and reduced the efficiency. Our further study demonstrated that CX45 overexpression or knockdown modulated the cell proliferation rate which was associated with the reprogramming efficiency. In conclusion, our data highlighted the critical role of CX45 in reprogramming and may increase the cell division rate and result in an accelerated kinetics of iPSCs production.
Sigma-Aldrich
Anti-Connexin 45 Antibody, near CT, cytoplasmic, clone 8A11.2, clone 8A11.2, Chemicon®, from mouse