Merck
  • Influence of Order within Nonpolar Monolayers on Hydrophobic Interactions.
Influence of Order within Nonpolar Monolayers on Hydrophobic Interactions.
Langmuir : the ACS journal of surfaces and colloids (2017-04-20)
Hongseung Yeon, Chenxuan Wang, Reid C Van Lehn, Nicholas L Abbott
ABSTRACT
We report an experimental investigation of the influence of molecular-level order (crystallinity) within nonpolar monolayers on hydrophobic interactions. The measurements were performed using gold film-supported monolayers formed from alkanethiols (CH
Sigma-Aldrich
1-Tetradecanethiol, ≥98.0% (GC)
Sigma-Aldrich
1-Decanethiol, 96%
Sigma-Aldrich
1-Octadecanethiol, 98%
Sigma-Aldrich
1-Hexanethiol, 99% (GC)
Sigma-Aldrich
1-Hexadecanethiol, ≥95.0% (GC)
Sigma-Aldrich
1-Octanethiol, ≥98.5%
Sigma-Aldrich
1-Butanethiol, 99%
Sigma-Aldrich
1-Dodecanethiol, ≥98%