Merck
  • Evaluation of efficient glucose release using sodium hydroxide and phosphoric acid as pretreating agents from the biomass of Sesbania grandiflora (L.) Pers.: A fast growing tree legume.
Evaluation of efficient glucose release using sodium hydroxide and phosphoric acid as pretreating agents from the biomass of Sesbania grandiflora (L.) Pers.: A fast growing tree legume.
Bioresource technology (2017-04-09)
Nitesh K Mund, Debabrata Dash, Chitta R Barik, Vaibhav V Goud, Lingaraj Sahoo, Prasannajit Mishra, Nihar R Nayak
ABSTRACT
Sesbania grandiflora (L.) Pers. is one of the fast growing tree legumes having the efficiency to produce around 50tha
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g