Merck
  • Terrein performs antitumor functions on esophageal cancer cells by inhibiting cell proliferation and synergistic interaction with cisplatin.
Terrein performs antitumor functions on esophageal cancer cells by inhibiting cell proliferation and synergistic interaction with cisplatin.
Oncology letters (2017-04-30)
Yanqing Wu, Yinghua Zhu, Shunrong Li, Minhua Zeng, Junjun Chu, Pengnan Hu, Jingjing Li, Qiannan Guo, Xiao-Bin Lv, Guofu Huang
ABSTRACT
Terrein is a bioactive fungal metabolite isolated from
Sigma-Aldrich
Terrein, from Aspergillus terreus, ≥98% (HPLC)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody