Merck
  • Effects of Antifouling Biocides on Molecular and Biochemical Defense System in the Gill of the Pacific Oyster Crassostrea gigas.
Effects of Antifouling Biocides on Molecular and Biochemical Defense System in the Gill of the Pacific Oyster Crassostrea gigas.
PloS one (2016-12-23)
Mi Seon Park, Young Dae Kim, Bo-Mi Kim, Youn-Jung Kim, Jang Kyun Kim, Jae-Sung Rhee
ABSTRACT
Antifouling biocides such as organotin compounds and their alternatives are potent toxicants in marine ecosystems. In this study, we employed several molecular and biochemical response systems of the Pacific oyster Crassostrea gigas to understand a potential mode of action of antifouling biocides (i.e. tributyltin (TBT), diuron and irgarol) after exposure to different concentrations (0.01, 0.1, and 1 μg L-1) for 96 h. As a result, all the three antifouling biocides strongly induced the antioxidant defense system. TBT reduced both enzymatic activity and mRNA expression of Na+/K+-ATPase and acetylcholinesterase (AChE). Lower levels of both Na+/K+-ATPase activity and AChE mRNA expression were observed in the diuron-exposed oysters compared to the control, while the irgarol treatment reduced only the transcriptional expression of AChE gene. We also analyzed transcript profile of heat shock protein (Hsp) superfamily in same experimental conditions. All antifouling biocides tested in this study significantly modulated mRNA expression of Hsp superfamily with strong induction of Hsp70 family. Taken together, overall results indicate that representative organotin TBT and alternatives have potential hazardous effects on the gill of C. gigas within relatively short time period. Our results also suggest that analyzing a series of molecular and biochemical parameters can be a way of understanding and uncovering the mode of action of emerging antifouling biocides. In particular, it was revealed that Pacific oysters have different sensitivities depend on the antifouling biocides.
Sigma-Aldrich
Glutathione Assay Kit, sufficient for 700 assays
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.
Ouabain, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glutathione Reductase Assay Kit, Sufficient for 100 colorimetric tests
Sigma-Aldrich
Catalase Assay Kit, sufficient for ≥100 tests enzymatic, determination of catalase activity in tissues and cells
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
Glutathione Peroxidase Cellular Activity Assay Kit, Sufficient for 100 colorimetric tests