Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Do K
Do K
British journal of pharmacology (2016-12-22)
Dmitry Tsvetkov, Mario Kaßmann, Jean-Yves Tano, Lan Chen, Johanna Schleifenbaum, Jakob Voelkl, Florian Lang, Yu Huang, Maik Gollasch
PMID
28000293
ABSTRACT
K We used Kcnq1 R-L3 produced similar concentration-dependent relaxations (EC We conclude that K
Product Description
C2615
Sigma-Aldrich
Chromanol 293B, ≥98% (HPLC), powder
Pricing
SML0524
Sigma-Aldrich
ML277, ≥98% (HPLC)
Pricing