Merck
  • Heparan Sulfate Domains Required for Fibroblast Growth Factor 1 and 2 Signaling through Fibroblast Growth Factor Receptor 1c.
Heparan Sulfate Domains Required for Fibroblast Growth Factor 1 and 2 Signaling through Fibroblast Growth Factor Receptor 1c.
The Journal of biological chemistry (2016-12-30)
Victor Schultz, Mathew Suflita, Xinyue Liu, Xing Zhang, Yanlei Yu, Lingyun Li, Dixy E Green, Yongmei Xu, Fuming Zhang, Paul L DeAngelis, Jian Liu, Robert J Linhardt
ABSTRACT
A small library of well defined heparan sulfate (HS) polysaccharides was chemoenzymatically synthesized and used for a detailed structure-activity study of fibroblast growth factor (FGF) 1 and FGF2 signaling through FGF receptor (FGFR) 1c. The HS polysaccharide tested contained both undersulfated (NA) domains and highly sulfated (NS) domains as well as very well defined non-reducing termini. This study examines differences in the HS selectivity of the positive canyons of the FGF1
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%
Sigma-Aldrich
Uridine 5′-diphospho-N-acetylglucosamine sodium salt, ≥98%