Merck
Multivalent Small-Molecule Pan-RAS Inhibitors.
Cell (2017-02-25)
Matthew E Welsch, Anna Kaplan, Jennifer M Chambers, Michael E Stokes, Pieter H Bos, Arie Zask, Yan Zhang, Marta Sanchez-Martin, Michael A Badgley, Christine S Huang, Timothy H Tran, Hemanth Akkiraju, Lewis M Brown, Renu Nandakumar, Serge Cremers, Wan Seok Yang, Liang Tong, Kenneth P Olive, Adolfo Ferrando, Brent R Stockwell
ABSTRACT
Design of small molecules that disrupt protein-protein interactions, including the interaction of RAS proteins and their effectors, may provide chemical probes and therapeutic agents. We describe here the synthesis and testing of potential small-molecule pan-RAS ligands, which were designed to interact with adjacent sites on the surface of oncogenic KRAS. One compound, termed 3144, was found to bind to RAS proteins using microscale thermophoresis, nuclear magnetic resonance spectroscopy, and isothermal titration calorimetry and to exhibit lethality in cells partially dependent on expression of RAS proteins. This compound was metabolically stable in liver microsomes and displayed anti-tumor activity in xenograft mouse cancer models. These findings suggest that pan-RAS inhibition may be an effective therapeutic strategy for some cancers and that structure-based design of small molecules targeting multiple adjacent sites to create multivalent inhibitors may be effective for some proteins.
Sigma-Aldrich
MISSION® esiRNA, targeting human HRAS
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Phosphatase, Alkaline−Agarose from calf intestine, ammonium sulfate suspension
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
RPMI 1640 Vitamins Solution (100×), liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Acridine, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
MISSION® esiRNA, targeting human NRAS
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS