Merck
  • YAP functions as a mechanotransducer in oligodendrocyte morphogenesis and maturation.
YAP functions as a mechanotransducer in oligodendrocyte morphogenesis and maturation.
Glia (2016-11-04)
Takeshi Shimizu, Yasuyuki Osanai, Kenji F Tanaka, Manabu Abe, Rie Natsume, Kenji Sakimura, Kazuhiro Ikenaka
ABSTRACT
Oligodendrocytes (OLs) are myelinating cells of the central nervous system. Recent studies have shown that mechanical factors influence various cell properties. Mechanical stimuli can be transduced into intracellular biochemical signals through mechanosensors and intracellular mechanotransducers, such as YAP. However, the molecular mechanisms underlying mechanical regulation of OLs by YAP remain unknown. We found that OL morphology and interactions between OLs and neuronal axons were affected by knocking down YAP. Mechanical stretching of OL precursor cells induced nuclear YAP accumulation and assembly of focal adhesion, key platforms for mechanotransduction. Shear stress decreased the number of OL processes, whereas a dominant-negative form of YAP suppressed these effects. To investigate the roles of YAP in postnatal OLs in vivo, we constructed a novel YAP knock-in mouse and found that in vivo overexpression of YAP widely affected OL maturation. These results indicate that YAP regulates OL morphology and maturation in response to mechanical factors. GLIA 2017;65:360-374.
Sigma-Aldrich
5-Fluoro-2′-deoxyuridine, thymidylate synthase inhibitor
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)