Merck
Alox5 Blockade Eradicates JAK2V617F-Induced Polycythemia Vera in Mice.
Cancer research (2016-10-28)
Yaoyu Chen, Yi Shan, Min Lu, Ngoc DeSouza, Zhiru Guo, Ronald Hoffman, Aibin Liang, Shaoguang Li
ABSTRACT
Myeloproliferative neoplasms such as polycythemia vera (PV), which are associated with the JAK mutation V617F, remain incurable despite progress in the use of JAK2 inhibitors for treatment of some of these diseases. In this study, we employed mice that undergo JAK2V617F-induced PV as a tool to explore new candidate targets for therapy. Our investigations focused on the lipid metabolic enzyme arachidonate 5-lipoxygenase (Alox5), which we found to be strongly upregulated by JAK2V617F in hematopoietic cells in vitro and in vivo Notably, genetic deletion of Alox5 or its inhibition in mice with a bioactive small-molecule inhibitor was sufficient to attenuate PV development. This therapeutic effect was associated with induction of a blockade in cell-cycle progression and also with apoptosis in PV cells. Genetic loss exerted an inhibitory effect on PV-initiating cells. Similarly, Alox5 inhibition was sufficient to suppress colony formation in human JAK2V617F-expressing CD34
Sigma-Aldrich
Leukotriene B4, ~100 μg/mL in ethanol, ≥97%