Merck
  • Dexrazoxane Shows No Protective Effect in the Acute Phase of Reperfusion during Myocardial Infarction in Pigs.
Dexrazoxane Shows No Protective Effect in the Acute Phase of Reperfusion during Myocardial Infarction in Pigs.
PloS one (2016-12-22)
Pranitha Kamat, Stijn Vandenberghe, Stephan Christen, Anjan K Bongoni, Bernhard Meier, Robert Rieben, Ahmed A Khattab
ABSTRACT
Calcium and iron overload participate in the mechanisms of ischemia/reperfusion (I/R) injury during myocardial infarction (MI). Calcium overload induces cardiomyocyte death by hypercontraction, while iron catalyses generation of reactive oxygen species (ROS). We therefore hypothesized that dexrazoxane, an intracellular metal chelator, would attenuate I/R injury. MI was induced in pigs by occlusion of the left anterior descending artery for 1 hour followed by 2 hours reperfusion. Thirty minutes before reperfusion either 5 mg/ml dexrazoxane (n = 5) or saline (n = 5) was infused intravenously. Myocardial necrosis as percentage of the area at ischemic risk was found to be similar in both groups (77.2 ± 18% for dexrazoxane and 76.4 ± 14% for saline group) as determined by triphenyl tetrazolium chloride staining of the ischemic myocardium. Also, serum levels of troponin-I were similar in both groups. A conductance catheter was used to measure left ventricular pressure and volume at all times. Markers for tissue damage due to ROS (HNE), endothelial cell activation (CD31) and inflammation (IgG, C3b/c, C5b9, MCP-1) were assessed on tissue and/or in serum. No significant differences were observed between the groups for the parameters analyzed. To conclude, in this clinically relevant model of early reperfusion after acute myocardial ischemia, dexrazoxane lacked attenuating effects on I/R injury as shown by the measured parameters.
Sigma-Aldrich
Dexrazoxane, ≥95% (HPLC)
Sigma-Aldrich
Anti-Complement 5b-9 Rabbit pAb, liquid, Calbiochem®