Merck
  • Expression and Clinical Significance of Serum Dipeptidyl Peptidase IV Chronic Obstructive Pulmonary Disease.
Expression and Clinical Significance of Serum Dipeptidyl Peptidase IV Chronic Obstructive Pulmonary Disease.
The American journal of the medical sciences (2016-03-20)
Xiao-Yue Chang, Yong Yang, Xiao-Qing Jia, Yuan Wang, Li-Na Peng, Xiao-Hong Ai, Cui-Ying Jiang, Jian-Hua Guo, Ting-Ting Wu
ABSTRACT
The purpose of this study is to explore the correlation between serum dipeptidyl peptidase IV (DPPIV) and chronic obstructive pulmonary disease (COPD) at its various disease states, analyze its applications in the prediction and diagnosis of COPD and test the possibility of DPPIV as the serologic marker for COPD screening. Samples from 74 patients (42 cases with acute exacerbation of COPD or acute exacerbation COPD (AECOPD) and 32 cases with stable COPD) and 29 control subjects were collected in this study. Those patients with AECOPD were classified as COPD remission group if their clinical symptoms relieved after nonintravenous or oral hormone therapy for 7 ± 3 days. DPPIV concentration was measured by enzyme-linked immunosorbent assay, and the difference in serum concentration of DPPIV was compared among different groups. The correlation between DPPIV concentration and age, sex or smoking history was analyzed, and the diagnostic value of DPPIV was evaluated by receiver-operating characteristic (ROC) curve analysis. Serum DPPIV concentration was significantly lower in all COPD groups as compared with that in healthy control group (P < 0.001). Serum DPPIV concentration in AECOPD group was increased after treatment (P < 0.001). There was no significant correlation between DPPIV concentration and age, sex or smoking history (P > 0.05). ROC analysis indicated that serum DPPIV concentration in all groups showed a good diagnostic accuracy, especially in stable COPD and AECOPD groups. The area under the ROC curve values were 0.901 and 0.906, respectively, with a high specificity of 0.931 for both groups and a high sensitivity of 0.75 for stable COPD and 0.875 for AECOPD. Serum DPPIV concentration in patients with COPD is decreased significantly, and there is no correlation between serum DPPIV concentration and sex or age. Serum DPPIV not only is an independent predictive factor, but also of high value as a good serologic marker for the diagnosis of COPD.