- Reaction of antibiotic sulfadiazine with manganese dioxide in aqueous phase: Kinetics, pathways and toxicity assessment.
Reaction of antibiotic sulfadiazine with manganese dioxide in aqueous phase: Kinetics, pathways and toxicity assessment.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2016-10-22)
Ji-Feng Yang, Li-Ming Yang, Guang-Guo Ying, Cheng-Bin Liu, Li-Ying Zheng, Sheng-Lian Luo
PMID27768526
ABSTRACT
Sulfonamide antibiotics are often detected in terrestrial and aquatic environment, but little is known about abiotic degradation of these antibiotics. In the present study, the degradation of the sulfonamide antibiotic sulfadiazine by a synthesized δ-MnO