Merck
  • USP19 deubiquitinates HDAC1/2 to regulate DNA damage repair and control chromosomal stability.
USP19 deubiquitinates HDAC1/2 to regulate DNA damage repair and control chromosomal stability.
Oncotarget (2016-08-16)
Min Wu, Hai-Qing Tu, Yan Chang, Bo Tan, Guang Wang, Jie Zhou, Li Wang, Rui Mu, Wei-Na Zhang
ABSTRACT
Excessive accumulation of DNA damage will generate chromosome stress, leading to various chromosome abnormalities such as chromatin bridge and result in genomic instability. Orchestra procession and regulation of DNA damage repair are vital for keeping genome stability. Despite of the key role of HDAC1/2 in double strand break (DSB) repair, the regulation for their mode of action is less well understood. In this study, we found that deubiquitination enzymes USP19 physically interacts with HDAC1/2 and specifically regulate their K63-linked ubiquitination, which might be crucial for regulation of HDAC1/2 activity in DNA damage repair. Notably, we found that USP19 trans-locate into nucleus upon IR irradiation and is indispensable for normally DNA damage response. In addition, we showed that USP19 play critical role in preventing anaphase bridge formation through regulating DNA damage repair process. Furthermore, the expression level of USP19 is commonly lower or deleted in several types of tumor. These results indicated that USP19 is a key factor in modulating DNA damage repair by targeting HDAC1/2 K63-linked ubiquitination, cells with deletion or decreased expression of USP19 might cause genome instability and even contribute to tumorigenesis.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)