Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Induced Neural Differentiation of MMP-2 Cleaved (RADA)
Induced Neural Differentiation of MMP-2 Cleaved (RADA)
Journal of controlled release : official journal of the Controlled Release Society (2016-11-05)
K Koss, C Tsui, L D Unsworth
PMID
27720765
ABSTRACT
(RADA)
Product Description
616044
Sigma-Aldrich
Fmoc-Ala-OH-2,3,3,3-d
4
, 98 atom % D
Pricing
233412
Sigma-Aldrich
Ethyl (hydroxyimino)cyanoacetate, 97%
Pricing