Merck
The antifolates.
Hematology/oncology clinics of North America (2012-04-24)
Michele Visentin, Rongbao Zhao, I David Goldman
ABSTRACT
This article focuses on the cellular, biochemical, and molecular pharmacology of antifolates and how a basic understanding of the mechanism of action of methotrexate, its cytotoxic determinants, mechanisms of resistance, and transport into and out of cells has led to the development of a new generation of antifolates, a process that continues in the laboratory and in the clinics. New approaches to folate-based cancer chemotherapy are described based on the targeted delivery of drugs to malignant cells.
Sigma-Aldrich
Aminopterin Ready Made Solution, 50 mg/mL in DMSO