Merck
Nanostructured materials for photon detection.
Nature nanotechnology (2010-05-18)
Gerasimos Konstantatos, Edward H Sargent
ABSTRACT
The detection of photons underpins imaging, spectroscopy, fibre-optic communications and time-gated distance measurements. Nanostructured materials are attractive for detection applications because they can be integrated with conventional silicon electronics and flexible, large-area substrates, and can be processed from the solution phase using established techniques such as spin casting, spray coating and layer-by-layer deposition. In addition, their performance has improved rapidly in recent years. Here we review progress in light sensing using nanostructured materials, focusing on solution-processed materials such as colloidal quantum dots and metal nanoparticles. These devices exhibit phenomena such as absorption of ultraviolet light, plasmonic enhancement of absorption, size-based spectral tuning, multiexciton generation, and charge carrier storage in surface and interface traps.
Sigma-Aldrich
PbS core-type quantum dots, oleic acid coated, fluorescence λem 900 nm, 10 mg/mL in toluene
Sigma-Aldrich
PbS core-type quantum dots, oleic acid coated, fluorescence λem 1500 nm, 10 mg/mL in toluene
Sigma-Aldrich
PbS core-type quantum dots, fluorescence λem 1600 nm, 10 mg/mL in toluene
Sigma-Aldrich
PbS core-type quantum dots, oleic acid coated, fluorescence λem 1100 nm, 10 mg/mL in toluene
Sigma-Aldrich
PbS core-type quantum dots, fluorescence λem 1000 nm, 10 mg/mL in toluene
Sigma-Aldrich
PbS core-type quantum dots, fluorescence λem 1400 nm, 10 mg/mL in toluene