Merck
  • Identification of novel γ-secretase-associated proteins in detergent-resistant membranes from brain.
Identification of novel γ-secretase-associated proteins in detergent-resistant membranes from brain.
The Journal of biological chemistry (2012-02-09)
Ji-Yeun Hur, Yasuhiro Teranishi, Takahiro Kihara, Natsuko Goto Yamamoto, Mitsuhiro Inoue, Waltteri Hosia, Masakazu Hashimoto, Bengt Winblad, Susanne Frykman, Lars O Tjernberg
ABSTRACT
In Alzheimer disease, oligomeric amyloid β-peptide (Aβ) species lead to synapse loss and neuronal death. γ-Secretase, the transmembrane protease complex that mediates the final catalytic step that liberates Aβ from its precursor protein (APP), has a multitude of substrates, and therapeutics aimed at reducing Aβ production should ideally be specific for APP cleavage. It has been shown that APP can be processed in lipid rafts, and γ-secretase-associated proteins can affect Aβ production. Here, we use a biotinylated inhibitor for affinity purification of γ-secretase and associated proteins and mass spectrometry for identification of the purified proteins, and we identify novel γ-secretase-associated proteins in detergent-resistant membranes from brain. Furthermore, we show by small interfering RNA-mediated knockdown of gene expression that a subset of the γ-secretase-associated proteins, in particular voltage-dependent anion channel 1 (VDAC1) and contactin-associated protein 1 (CNTNAP1), reduced Aβ production (Aβ40 and Aβ42) by around 70%, whereas knockdown of presenilin 1, one of the essential γ-secretase complex components, reduced Aβ production by 50%. Importantly, these proteins had a less pronounced effect on Notch processing. We conclude that VDAC1 and CNTNAP1 associate with γ-secretase in detergent-resistant membranes and affect APP processing and suggest that molecules that interfere with this interaction could be of therapeutic use for Alzheimer disease.
Sigma-Aldrich
Anti-Nicastrin Antibody, ascites fluid, clone 9C3, Chemicon®