Merck
  • SUVR2 is involved in transcriptional gene silencing by associating with SNF2-related chromatin-remodeling proteins in Arabidopsis.
SUVR2 is involved in transcriptional gene silencing by associating with SNF2-related chromatin-remodeling proteins in Arabidopsis.
Cell research (2014-11-26)
Yong-Feng Han, Kun Dou, Ze-Yang Ma, Su-Wei Zhang, Huan-Wei Huang, Lin Li, Tao Cai, She Chen, Jian-Kang Zhu, Xin-Jian He
ABSTRACT
The SU(VAR)3-9-like histone methyltransferases usually catalyze repressive histone H3K9 methylation and are involved in transcriptional gene silencing in eukaryotic organisms. We identified a putative SU(VAR)3-9-like histone methyltransferase SUVR2 by a forward genetic screen and demonstrated that it is involved in transcriptional gene silencing at genomic loci targeted by RNA-directed DNA methylation (RdDM). We found that SUVR2 has no histone methyltransferase activity and the conserved catalytic sites of SUVR2 are dispensable for the function of SUVR2 in transcriptional silencing. SUVR2 forms a complex with its close homolog SUVR1 and associate with three previously uncharacterized SNF2-related chromatin-remodeling proteins CHR19, CHR27, and CHR28. SUVR2 was previously thought to be a component in the RdDM pathway. We demonstrated that SUVR2 contributes to transcriptional gene silencing not only at a subset of RdDM target loci but also at many RdDM-independent target loci. Our study suggests that the involvement of SUVR2 in transcriptional gene silencing is related to nucleosome positioning mediated by its associated chromatin-remodeling proteins.
Sigma-Aldrich
PerfectHyb Plus Hybridization Buffer, for Northern and Southern blotting, solution
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys9) - ChIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
ProteoSilver Silver Stain Kit