Merck
  • Suppression of ROS Production by Exendin-4 in PSC Attenuates the High Glucose-Induced Islet Fibrosis.
Suppression of ROS Production by Exendin-4 in PSC Attenuates the High Glucose-Induced Islet Fibrosis.
PloS one (2016-12-16)
Ji-Won Kim, Shin-Young Park, Young-Hye You, Dong-Sik Ham, Seung-Hwan Lee, Hae Kyung Yang, In-Kyung Jeong, Seung-Hyun Ko, Kun-Ho Yoon
ABSTRACT
Pancreatic stellate cells (PSCs) play a major role to fibrotic islet destruction observed in diabetic patients and animal model of diabetes. Exendin-4 (Ex-4) is a potent insulinotropic agent and has been approved for the treatment of type 2 diabetes. However, there have been no reports demonstrating the effects of Ex-4 on pancreatic islet fibrosis. In this study, Ex-4 treatment clearly attenuated fibrotic islet destruction and improved glucose tolerance and islet survival. GLP-1 receptor expression was upregulated during activation and proliferation of PSCs by hyperglycemia. The activation of PKA pathway by Ex-4 plays a role in ROS production and angiotensin II (Ang II) production. Exposure to high glucose stimulated ERK activation and Ang II-TGF- β1 production in PSCs. Interestingly, Ex-4 significantly reduced Ang II and TGF-β1 production by inhibition of ROS production but not ERK phosphorylation. Ex-4 may be useful not only as an anti-diabetic agent but also as an anti-fibrotic agent in type 2 diabetes due to its ability to inhibit PSC activation and proliferation and improve islet fibrosis in OLETF rats.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Exendin-4, ≥97%