Merck
  • Regional differences in rat vaginal smooth muscle contractility and morphology.
Regional differences in rat vaginal smooth muscle contractility and morphology.
Reproductive sciences (Thousand Oaks, Calif.) (2013-01-10)
Laura C Skoczylas, Zegbeh Jallah, Yoshio Sugino, Suzan E Stein, Andrew Feola, Naoki Yoshimura, Pamela Moalli
ABSTRACT
The objective of this study was to define the regional differences in rat vaginal smooth muscle contractility and morphology. We evaluated circumferential segments from the proximal, middle, and distal rat vagina (n = 21) in vitro. Contractile responses to carbachol, phenylephrine, potassium chloride, and electrical field stimulation (EFS) were measured. Immunohistochemical analyses were also performed. The dose-response curves for carbachol- and phenylephrine-dependent contractions were different in the distal (P = .05, P = .04) compared to the proximal/middle regions. Adjusted for region-dependent changes in contractility, the distal vagina generated lower force in response to carbachol and higher force in response to phenylephrine. There was less force with increasing EFS frequency in the distal (P = .03), compared to the proximal/middle regions. Cholinergic versus adrenergic nerves were more frequent in the proximal region (P = .03). In summary, the results indicate that functional and morphological differences in smooth muscle and nerve fibers of the distal versus proximal/middle regions of the vagina exist.