Merck
Overexpression of neuritin in gastric cancer.
Oncology letters (2016-01-21)
Ming Yuan, Yongjun Li, Chen Zhong, Yongkang Li, Jianhua Niu, Jianping Gong
ABSTRACT
The aim of the present study was to investigate the expression of neuritin in gastric cancer tissues, in order to explore the association between the expression of neuritin and the occurrence and development of gastric cancer. Tissue specimens were collected from 58 patients with gastric cancer. Immunohistochemistry, western blot analysis and reverse transcription-polymerase chain reaction (RT-PCR) were used to determine the expression of neuritin in the gastric cancer and corresponding adjacent normal gastric tissues. The expression rate of neuritin in gastric cancer tissues was 96.55% (56/58), demonstrating no statistically significant difference from the expression rate in the adjacent normal tissues (94.83%) (P>0.05). However, the rate of strong neuritin expression in gastric cancer tissues (82.76%) was significantly increased compared with the rate in the adjacent normal tissues (15.52%) (P<0.05). Neuritin expression exhibited no correlation with the gender or age of patients, tumor-node-metastasis staging, tumor depth, presence of lymph node metastasis, histological or pathological type of the tumor or presence of distant metastasis (P>0.05). As determined by RT-PCR and western blot analysis, the mRNA expression of neuritin in gastric cancer tissues was markedly increased compared with the expression in the adjacent normal tissues. In conclusion, neuritin is highly expressed in gastric cancer tissues, suggesting that neuritin may act as a novel potential target for the treatment of gastric cancer.