Merck
  • Antibody production using a ciliate generates unusual antibody glycoforms displaying enhanced cell-killing activity.
Antibody production using a ciliate generates unusual antibody glycoforms displaying enhanced cell-killing activity.
mAbs (2016-10-30)
Jenny Calow, Anna-Janina Behrens, Sonja Mader, Ulrike Bockau, Weston B Struwe, David J Harvey, Kai U Cormann, Marc M Nowaczyk, Karin Loser, Daniel Schinor, Marcus W W Hartmann, Max Crispin
ABSTRACT
Antibody glycosylation is a key parameter in the optimization of antibody therapeutics. Here, we describe the production of the anti-cancer monoclonal antibody rituximab in the unicellular ciliate, Tetrahymena thermophila. The resulting antibody demonstrated enhanced antibody-dependent cell-mediated cytotoxicity, which we attribute to unusual N-linked glycosylation. Detailed chromatographic and mass spectrometric analysis revealed afucosylated, oligomannose-type glycans, which, as a whole, displayed isomeric structures that deviate from the typical human counterparts, but whose branches were equivalent to fragments of metabolic intermediates observed in human glycoproteins. From the analysis of deposited crystal structures, we predict that the ciliate glycans adopt protein-carbohydrate interactions with the Fc domain that closely mimic those of native complex-type glycans. In addition, terminal glucose structures were identified that match biosynthetic precursors of human glycosylation. Our results suggest that ciliate-based expression systems offer a route to large-scale production of monoclonal antibodies exhibiting glycosylation that imparts enhanced cell killing activity.