Merck
The Microprocessor controls the activity of mammalian retrotransposons.
Nature structural & molecular biology (2013-09-03)
Sara R Heras, Sara Macias, Mireya Plass, Noemí Fernandez, David Cano, Eduardo Eyras, José L Garcia-Perez, Javier F Cáceres
ABSTRACT
More than half of the human genome is made of transposable elements whose ongoing mobilization is a driving force in genetic diversity; however, little is known about how the host regulates their activity. Here, we show that the Microprocessor (Drosha-DGCR8), which is required for microRNA biogenesis, also recognizes and binds RNAs derived from human long interspersed element 1 (LINE-1), Alu and SVA retrotransposons. Expression analyses demonstrate that cells lacking a functional Microprocessor accumulate LINE-1 mRNA and encoded proteins. Furthermore, we show that structured regions of the LINE-1 mRNA can be cleaved in vitro by Drosha. Additionally, we used a cell culture-based assay to show that the Microprocessor negatively regulates LINE-1 and Alu retrotransposition in vivo. Altogether, these data reveal a new role for the Microprocessor as a post-transcriptional repressor of mammalian retrotransposons and a defender of human genome integrity.
Product Description
Millipore
T7•Tag® Antibody Agarose