Merck
  • A novel mitofusin 2 mutation causes canine fetal-onset neuroaxonal dystrophy.
A novel mitofusin 2 mutation causes canine fetal-onset neuroaxonal dystrophy.
Neurogenetics (2011-06-07)
John C Fyfe, Rabá A Al-Tamimi, Junlong Liu, Alejandro A Schäffer, Richa Agarwala, Paula S Henthorn
ABSTRACT
We recently reported autosomal recessive fetal-onset neuroaxonal dystrophy (FNAD) in a large family of dogs that is not caused by mutation in the PLA2G6 locus (Fyfe et al., J Comp Neurol 518:3771-3784, 2010). Here, we report a genome-wide linkage analysis using 333 microsatellite markers to map canine FNAD to the telomeric end of chromosome 2. The interval of zero recombination was refined by single-nucleotide polymorphism (SNP) haplotype analysis to ~200 kb, and the included genes were sequenced. We found a homozygous 3-nucleotide deletion in exon 14 of mitofusin 2 (MFN2), predicting loss of a glutamate residue at position 539 in the protein of affected dogs. RT-PCR demonstrated near normal expression of the mutant mRNA, but MFN2 expression was undetectable to very low on western blots of affected dog brainstem, cerebrum, kidney, and cultured fibroblasts and by immunohistochemistry on brainstem sections. MFN2 is a multifunctional, membrane-bound GTPase of mitochondria and endoplasmic reticulum most commonly associated with human Charcot-Marie-Tooth disease type 2A2. The canine disorder extends the range of MFN2-associated phenotypes and suggests MFN2 as a candidate gene for rare cases of human FNAD.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution