Merck
  • Deletion of the tyrosine phosphatase Shp2 in Sertoli cells causes infertility in mice.
Deletion of the tyrosine phosphatase Shp2 in Sertoli cells causes infertility in mice.
Scientific reports (2015-08-13)
Xiaopeng Hu, Zhenzhou Tang, Yang Li, Wensheng Liu, Shuang Zhang, Bingyan Wang, Yingpu Tian, Yinan Zhao, Hao Ran, Wenjie Liu, Gen-Sheng Feng, Jianwei Shuai, Haibin Wang, Zhongxian Lu
ABSTRACT
The male's ability to reproduce is completely dependent on Sertoli cells. However, the mechanisms governing the functional integrity of Sertoli cells have remained largely unexplored. Here, we demonstrate that deletion of Shp2 in Sertoli cells results in infertility in mice. In Shp2 knockout mice (SCSKO), a normal population of Sertoli cells was observed, but the blood-testis barrier (BTB) was not formed. Shp2 ablation initiated the untimely and excessive differentiation of spermatogonial stem cells (SSCs) by disturbing the expression of paracrine factors. As a consequence, the process of spermatogenesis was disrupted, and the germ cells were depleted. Furthermore, Shp2 deletion impaired the cell junctions of the primary Sertoli cells and failed to support the clonal formation of SSCs co-cultured with SCSKO Sertoli cells. As expected, Shp2 restoration largely restores the cell junctions of the primary Sertoli cells and the clonal formation of SSCs. To identify the underlying mechanism, we further demonstrated that the absence of Shp2 suppressed Erk phosphorylation, and thus, the expression of follicle-stimulating hormone (FSH)- and testosterone-induced target genes. These results collectively suggest that Shp2 is a critical signaling protein that is required to maintain Sertoli cell function and could serve as a novel target for male infertility therapies.