Merck
  • Cross-talk between PRMT1-mediated methylation and ubiquitylation on RBM15 controls RNA splicing.
Cross-talk between PRMT1-mediated methylation and ubiquitylation on RBM15 controls RNA splicing.
eLife (2015-11-18)
Li Zhang, Ngoc-Tung Tran, Hairui Su, Rui Wang, Yuheng Lu, Haiping Tang, Sayura Aoyagi, Ailan Guo, Alireza Khodadadi-Jamayran, Dewang Zhou, Kun Qian, Todd Hricik, Jocelyn Côté, Xiaosi Han, Wenping Zhou, Suparna Laha, Omar Abdel-Wahab, Ross L Levine, Glen Raffel, Yanyan Liu, Dongquan Chen, Haitao Li, Tim Townes, Hengbin Wang, Haiteng Deng, Y George Zheng, Christina Leslie, Minkui Luo, Xinyang Zhao
ABSTRACT
RBM15, an RNA binding protein, determines cell-fate specification of many tissues including blood. We demonstrate that RBM15 is methylated by protein arginine methyltransferase 1 (PRMT1) at residue R578, leading to its degradation via ubiquitylation by an E3 ligase (CNOT4). Overexpression of PRMT1 in acute megakaryocytic leukemia cell lines blocks megakaryocyte terminal differentiation by downregulation of RBM15 protein level. Restoring RBM15 protein level rescues megakaryocyte terminal differentiation blocked by PRMT1 overexpression. At the molecular level, RBM15 binds to pre-messenger RNA intronic regions of genes important for megakaryopoiesis such as GATA1, RUNX1, TAL1 and c-MPL. Furthermore, preferential binding of RBM15 to specific intronic regions recruits the splicing factor SF3B1 to the same sites for alternative splicing. Therefore, PRMT1 regulates alternative RNA splicing via reducing RBM15 protein concentration. Targeting PRMT1 may be a curative therapy to restore megakaryocyte differentiation for acute megakaryocytic leukemia.
Sigma-Aldrich
Anti-PRMT1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)