Merck
  • Proximity biotinylation and affinity purification are complementary approaches for the interactome mapping of chromatin-associated protein complexes.
Proximity biotinylation and affinity purification are complementary approaches for the interactome mapping of chromatin-associated protein complexes.
Journal of proteomics (2014-10-05)
Jean-Philippe Lambert, Monika Tucholska, Christopher Go, James D R Knight, Anne-Claude Gingras
ABSTRACT
Mapping protein-protein interactions for chromatin-associated proteins remains challenging. Here we explore the use of BioID, a proximity biotinylation approach in which a mutated biotin ligase (BirA*) is fused to a bait of interest, allowing for the local activation of biotin and subsequent biotinylation of proteins in the bait vicinity. BioID allowed for successful interactome mapping of core histones and members of the mediator complex. We explored the background signal produced by the BioID approach and found that using distinct types of controls increased the stringency of our statistical analysis with SAINTexpress. A direct comparison of BioID with our AP-MS protocol optimized for chromatin-associated protein complexes revealed that the approaches identified few shared interaction partners and enriched for distinct biological processes; yet, both approaches permitted the recovery of biologically meaningful interactions. While no clear bias could be observed for either technique toward protein complexes of particular functions, BioID allowed for the purification of proteins of lower cellular abundance. Finally, we were able to identify a strong association of MED4 with the centrosome by BioID and validated this finding by immunofluorescence. In summary, BioID complements AP-MS for the study of chromatin-associated protein complexes. This manuscript describes the application of BioID, a proximity biotinylation approach, to chromatin-associated proteins, namely core histones and members of the mediator complex. We observed that BioID was successful at identifying known interaction partners for the baits tested, but also allowed novel putative interaction partners to be identified. By performing a detailed comparison of BioID versus a standard method for interactome mapping (affinity purification coupled to mass spectrometry, AP-MS), we show that the approaches were complementary, allowing for purification of different interaction partners. These interaction partners were different in the biological processes they are associated with, but also in their abundance. BioID represents a significant technical development in the field of chromatin research by expanding the search space for interactome mapping beyond what is possible with AP-MS. This article is part of a Special Issue entitled: Protein dynamics in health and disease. Guest Editors: Pierre Thibault and Anne-Claude Gingras.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H3 (Lys27) Antibody, serum, Upstate®