Merck
  • Endosome-mediated autophagy: an unconventional MIIC-driven autophagic pathway operational in dendritic cells.
Endosome-mediated autophagy: an unconventional MIIC-driven autophagic pathway operational in dendritic cells.
Autophagy (2013-03-14)
Vangelis Kondylis, Hezder E van Nispen Tot Pannerden, Suzanne van Dijk, Toine Ten Broeke, Richard Wubbolts, Willie J Geerts, Cor Seinen, Tuna Mutis, Harry F G Heijnen
ABSTRACT
Activation of TLR signaling has been shown to induce autophagy in antigen-presenting cells (APCs). Using high-resolution microscopy approaches, we show that in LPS-stimulated dendritic cells (DCs), autophagosomes emerge from MHC class II compartments (MIICs) and harbor both the molecular machinery for antigen processing and the autophagosome markers LC3 and ATG16L1. This ENdosome-Mediated Autophagy (ENMA) appears to be the major type of autophagy in DCs, as similar structures were observed upon established autophagy-inducing conditions (nutrient deprivation, rapamycin) and under basal conditions in the presence of bafilomycin A1. Autophagosome formation was not significantly affected in DCs expressing ATG4B (C74A) mutant and atg4b (-/-) bone marrow DCs, but the degradation of the autophagy substrate SQSTM1/p62 was largely impaired. Furthermore, we demonstrate that the previously described DC aggresome-like LPS-induced structures (DALIS) contain vesicular membranes, and in addition to SQSTM1 and ubiquitin, they are positive for LC3. LC3 localization on DALIS is independent of its lipidation. MIIC-driven autophagosomes preferentially engulf the LPS-induced SQSTM1-positive DALIS, which become later degraded in autolysosomes. DALIS-associated membranes also contain ATG16L1, ATG9 and the Q-SNARE VTI1B, suggesting that they may represent (at least in part) a membrane reservoir for autophagosome expansion. We propose that ENMA constitutes an unconventional, APC-specific type of autophagy, which mediates the processing and presentation of cytosolic antigens by MHC class II machinery, and/or the selective clearance of toxic by-products of elevated ROS/RNS production in activated DCs, thereby promoting their survival.
Sigma-Aldrich
Anti-KDEL Mouse mAb (10C3), liquid, clone 10C3, Calbiochem®