Merck
  • Identification of regulators of the innate immune response to cytosolic DNA and retroviral infection by an integrative approach.
Identification of regulators of the innate immune response to cytosolic DNA and retroviral infection by an integrative approach.
Nature immunology (2012-12-25)
Mark N Lee, Matthew Roy, Shao-En Ong, Philipp Mertins, Alexandra-Chloé Villani, Weibo Li, Farokh Dotiwala, Jayita Sen, John G Doench, Megan H Orzalli, Igor Kramnik, David M Knipe, Judy Lieberman, Steven A Carr, Nir Hacohen
ABSTRACT
The innate immune system senses viral DNA that enters mammalian cells, or in aberrant situations self-DNA, and triggers type I interferon production. Here we present an integrative approach that combines quantitative proteomics, genomics and small molecule perturbations to identify genes involved in this pathway. We silenced 809 candidate genes, measured the response to dsDNA and connected resulting hits with the known signaling network. We identified ABCF1 as a critical protein that associates with dsDNA and the DNA-sensing components HMGB2 and IFI204. We also found that CDC37 regulates the stability of the signaling molecule TBK1 and that chemical inhibition of the CDC37-HSP90 interaction and several other pathway regulators potently modulates the innate immune response to DNA and retroviral infection.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid