Merck
A protease-independent function for SPPL3 in NFAT activation.
Molecular and cellular biology (2014-11-12)
Stefanie L Makowski, Zhaoquan Wang, Joel L Pomerantz
ABSTRACT
The signal peptide peptidase (SPP)-related intramembrane aspartyl proteases are a homologous group of polytopic membrane proteins, some of which function in innate or adaptive immunity by cleaving proteins involved in antigen presentation or intracellular signaling. Signal peptide peptidase-like 3 (SPPL3) is a poorly characterized endoplasmic reticulum (ER)-localized member of this family, with no validated cellular substrates. We report here the isolation of SPPL3 in a screen for activators of NFAT, a transcription factor that controls lymphocyte development and function. We find that SPPL3 is required downstream of T cell receptor engagement for maximal Ca(2+) influx and NFAT activation. Surprisingly, the proteolytic activity of SPPL3 is not required for its role in this pathway. SPPL3 enhances the signal-induced association of stromal interaction molecule 1 (STIM1) and Orai1 and is even required for the full activity of constitutively active STIM1 variants that bind Orai1 independently of ER Ca(2+) release. SPPL3 associates with STIM1 through at least two independent domains, the transmembrane region and the CRAC activation domain (CAD), and can promote the association of the STIM1 CAD with Orai1. Our results assign a function in lymphocyte signaling to SPPL3 and highlight the emerging importance of nonproteolytic functions for members of the intramembrane aspartyl protease family.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Anti-Orai1 Antibody, Intracellular, Chemicon®, from rabbit