Merck
  • The sirtuin SIRT6 regulates stress granule formation in C. elegans and mammals.
The sirtuin SIRT6 regulates stress granule formation in C. elegans and mammals.
Journal of cell science (2013-09-10)
Monika Jedrusik-Bode, Maja Studencka, Christian Smolka, Tobias Baumann, Henning Schmidt, Jan Kampf, Franziska Paap, Sophie Martin, Jamal Tazi, Kristian M Müller, Marcus Krüger, Thomas Braun, Eva Bober
ABSTRACT
SIRT6 is a NAD(+)-dependent deacetylase that modulates chromatin structure and safeguards genomic stability. Until now, SIRT6 has been assigned to the nucleus and only nuclear targets of SIRT6 are known. Here, we demonstrate that in response to stress, C. elegans SIR-2.4 and its mammalian orthologue SIRT6 localize to cytoplasmic stress granules, interact with various stress granule components and induce their assembly. Loss of SIRT6 or inhibition of its catalytic activity in mouse embryonic fibroblasts impairs stress granule formation and delays disassembly during recovery, whereas deficiency of SIR-2.4 diminishes maintenance of P granules and decreases survival of C. elegans under stress conditions. Our findings uncover a novel, evolutionary conserved function of SIRT6 in the maintenance of stress granules in response to stress.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)