Merck
  • Creation and characterization of BAC-transgenic mice with physiological overexpression of epitope-tagged RCAN1 (DSCR1).
Creation and characterization of BAC-transgenic mice with physiological overexpression of epitope-tagged RCAN1 (DSCR1).
Mammalian genome : official journal of the International Mammalian Genome Society (2012-10-26)
Luzhou Xing, Martha Salas, Hong Zhang, Julia Gittler, Thomas Ludwig, Chyuan-Sheng Lin, Vundavalli V Murty, Wayne Silverman, Ottavio Arancio, Benjamin Tycko
ABSTRACT
The chromosome 21 gene RCAN1, encoding a modulator of the calcineurin (CaN) phosphatase, is a candidate gene for contributing to cognitive disability in people with Down syndrome (DS; trisomy 21). To develop a physiologically relevant model for studying the biochemistry of RCAN1 and its contribution to DS, we generated bacterial artificial chromosome-transgenic (BAC-Tg) mouse lines containing the human RCAN1 gene with a C-terminal HA-FLAG epitope tag incorporated by recombineering. The BAC-Tg was expressed at levels only moderately higher than the native Rcan1 gene: approximately 1.5-fold in RCAN1 (BAC-Tg1) and twofold in RCAN1 (BAC-Tg2). Affinity purification of the RCAN1 protein complex from brains of these mice revealed a core complex of RCAN1 with CaN, glycogen synthase kinase 3-beta (Gsk3b), and calmodulin, with substoichiometric components, including LOC73419. The BAC-Tg mice are fully viable, but long-term synaptic potentiation is impaired in proportion to BAC-Tg dosage in hippocampal brain slices from these mice. RCAN1 can act as a tumor suppressor in some systems, but we found that the RCAN1 BAC-Tg did not reduce mammary cancer growth when present at a low copy number in Tp53;WAP-Cre mice. This work establishes a useful mouse model for investigating the biochemistry and dose-dependent functions of the RCAN1 protein in vivo.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution