Merck
  • PTEN regulates retinal interneuron morphogenesis and synaptic layer formation.
PTEN regulates retinal interneuron morphogenesis and synaptic layer formation.
Molecular and cellular neurosciences (2011-12-14)
Kiyo Sakagami, Bryan Chen, Steven Nusinowitz, Hong Wu, Xian-Jie Yang
ABSTRACT
The lipid phosphatase PTEN is a critical negative regulator of extracellular signal-induced PI3K activities, yet the roles of PTEN in the neural retina remain poorly understood. Here, we investigate the function of PTEN during retinal development. Deletion of Pten at the onset of neurogenesis in retinal progenitors results in the reduction of retinal ganglion cells and rod photoreceptors, but increased Müller glial genesis. In addition, PTEN deficiency leads to elevated phosphorylation of Akt, especially in the developing inner plexiform layer, where high levels of PTEN are normally expressed. In Pten mutant retinas, various subtypes of amacrine cells show severe dendritic overgrowth, causing specific expansion of the inner plexiform layer. However, the outer plexiform layer remains relatively undisturbed in the Pten deficient retina. Physiological analysis detects reduced rod function and augmented oscillatory potentials originating from amacrine cells in Pten mutants. Furthermore, deleting Pten or elevating Akt activity in individual amacrine cells is sufficient to disrupt dendritic arborization, indicating that Pten activity is required cell autonomously to control neuronal morphology. Moreover, inhibiting endogenous Akt activity attenuates inner plexiform layer formation in vitro. Together, these findings demonstrate that suppression of PI3K/Akt signaling by PTEN is crucial for proper neuronal differentiation and normal retinal network formation.
Sigma-Aldrich
Anti-Prox 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Chx10 Antibody, CT, Chemicon®, from sheep
Sigma-Aldrich
Anti-Brn-3a Antibody, POU-domain protein, clone 5A3.2, culture supernatant, clone 5A3.2, Chemicon®
Sigma-Aldrich
Anti-Glutamine Synthetase Antibody, clone GS-6, clone GS-6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat