Merck
  • Angiotensin II enhances AT1-Nox1 binding and stimulates arterial smooth muscle cell migration and proliferation through AT1, Nox1, and interleukin-18.
Angiotensin II enhances AT1-Nox1 binding and stimulates arterial smooth muscle cell migration and proliferation through AT1, Nox1, and interleukin-18.
American journal of physiology. Heart and circulatory physiology (2012-05-29)
Anthony J Valente, Tadashi Yoshida, Subramanyam N Murthy, Siva S V P Sakamuri, Masato Katsuyama, Robert A Clark, Patrice Delafontaine, Bysani Chandrasekar
ABSTRACT
The redox-sensitive transcription factors NF-κB and activator protein-1 (AP-1) are critical mediators of ANG II signaling. The promitogenic and promigratory factor interleukin (IL)-18 is an NF-κB- and AP-1-responsive gene. Therefore, we investigated whether ANG II-mediated smooth muscle cell (SMC) migration and proliferation involve IL-18. ANG II induced rat carotid artery SMC migration and proliferation and IL-18 and metalloproteinase (MMP)-9 expression via ANG II type 1 (AT(1)) receptor. ANG II-induced superoxide generation, NF-κB and AP-1 activation, and IL-18 and MMP-9 induction were all markedly attenuated by losartan, diphenyleneiodonium chloride (DPI), and Nox1 knockdown. Similar to ANG II, addition of IL-18 also induced superoxide generation, activated NF-κB and AP-1, and stimulated SMC migration and proliferation, in part via Nox1, and both ANG II and IL-18 induced NOX1 transcription in an AP-1-dependent manner. AT(1) physically associates with Nox1 in SMC, and ANG II enhanced this binding. Interestingly, exogenous IL-18 neither induced AT(1) binding to Nox1 nor enhanced the ANG II-induced increase in AT(1)/Nox1 binding. Importantly, IL-18 knockdown, or pretreatment with IL-18 neutralizing antibodies, or IL-18 binding protein, all attenuated the migratory and mitogenic effects of ANG II. Continuous infusion of ANG II for 7 days induced carotid artery hyperplasia in rats via AT(1) and was associated with increased AT(1)/Nox1 binding (despite lower AT(1) levels); increased DPI-inhibitable superoxide production; increased phospho-IKKβ, JNK, p65, and c-Jun; and induction of IL-18 and MMP-9 in endothelium-denuded carotid arteries. These results indicate that IL-18 amplifies the ANG II-induced, redox-dependent inflammatory cascades by activating similar promitogenic and promigratory signal transduction pathways. The ANG II/Nox1/IL-18 pathway may be critical in hyperplastic vascular diseases, including atherosclerosis and restenosis.