Merck
  • Simplifying and expanding analytical capabilities for various classes of doping agents by means of direct urine injection high performance liquid chromatography high resolution/high accuracy mass spectrometry.
Simplifying and expanding analytical capabilities for various classes of doping agents by means of direct urine injection high performance liquid chromatography high resolution/high accuracy mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2016-10-04)
Christian Görgens, Sven Guddat, Andreas Thomas, Philipp Wachsmuth, Anne-Katrin Orlovius, Gerd Sigmund, Mario Thevis, Wilhelm Schänzer
ABSTRACT
So far, in sports drug testing compounds of different classes are processed and measured using different screening procedures. The constantly increasing number of samples in doping analysis, as well as the large number of substances with doping related, pharmacological effects require the development of even more powerful assays than those already employed in sports drug testing, indispensably with reduced sample preparation procedures. The analysis of native urine samples after direct injection provides a promising analytical approach, which thereby possesses a broad applicability to many different compounds and their metabolites, without a time-consuming sample preparation. In this study, a novel multi-target approach based on liquid chromatography and high resolution/high accuracy mass spectrometry is presented to screen for more than 200 analytes of various classes of doping agents far below the required detection limits in sports drug testing. Here, classic groups of drugs as diuretics, stimulants, β
Sigma-Aldrich
Dorzolamide, ≥98% (HPLC)
Sigma-Aldrich
2-Aminoheptane, 99%
Sigma-Aldrich
(1S,2S)-(+)-Pseudoephedrine, 98%