Merck
  • Arginine modified polymeric micelles as a novel drug delivery system with enhanced endocytosis efficiency.
Arginine modified polymeric micelles as a novel drug delivery system with enhanced endocytosis efficiency.
Colloids and surfaces. B, Biointerfaces (2016-11-05)
Song Luo, Yixin Zhang, Jun Cao, Bin He, Sai Li
ABSTRACT
A new drug delivery system with improved endocytosis efficiency based on arginine modified poly(ethylene glycol)-b-poly(ε-caprolactone) (PEG-PCL) diblock copolymers was developed successfully. The structures of amphiphilic copolymers were verified by proton Nuclear Magnetic Resonance (
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Supelco
3,5,6-Trichloro-2-pyridinol, PESTANAL®, analytical standard
Sigma-Aldrich
1-Pyrenemethanol, 98%
Sigma-Aldrich
Boc-Arg(Pbf)-OH, ≥98.0% (TLC)