- Localized force application reveals mechanically sensitive domains of Piezo1.
Localized force application reveals mechanically sensitive domains of Piezo1.
Nature communications (2016-10-04)
Jason Wu, Raman Goyal, Jörg Grandl
PMID27694883
ABSTRACT
Piezos are mechanically activated ion channels that function as sensors of touch and pressure in various cell types. However, the precise mechanism and structures mediating mechanical activation and subsequent inactivation have not yet been identified. Here we use magnetic nanoparticles as localized transducers of mechanical force in combination with pressure-clamp electrophysiology to identify mechanically sensitive domains important for activation and inactivation.