Merck
  • Silver sulfadiazine loaded chitosan/chondroitin sulfate films for a potential wound dressing application
Silver sulfadiazine loaded chitosan/chondroitin sulfate films for a potential wound dressing application
Materials Science & Engineering. C, Materials For Biological Applications (2013)
Fajardo AR, et al.