Merck
Catalytic asymmetric formation of beta-sultams.
Organic letters (2007-04-24)
Marian Zajac, René Peters
ABSTRACT
beta-Sultams, highly strained sulfonyl analogues of beta-lactams, were prepared enantio- and diastereoselectively by tertiary amine catalyzed [2 + 2] cycloaddition reactions. The title compounds are practical precursors of highly enantioenriched biologically interesting beta-aminosulfonyl derivatives.
Sigma-Aldrich
3-Chloropropanesulfonyl chloride, 98%
Sigma-Aldrich
Sulfuryl chloride solution, 1.0 M in methylene chloride