- Asymmetric catalytic Mannich reactions catalyzed by urea derivatives: enantioselective synthesis of beta-aryl-beta-amino acids.
Asymmetric catalytic Mannich reactions catalyzed by urea derivatives: enantioselective synthesis of beta-aryl-beta-amino acids.
Journal of the American Chemical Society (2002-10-31)
Anna G Wenzel, Eric N Jacobsen
PMID12405820
ABSTRACT
Highly enantioselective addition reactions between silyl ketene acetals and N-Boc aldimines are catalyzed by the thiourea-based catalyst 1c. Extraordinary scope is observed in this methodology with regard to the imine substrate, with aryl and heteroaromatic derivatives generally affording nearly quantitative yields of beta-amino ester product in up to 98% enantioselectivity.