Merck
  • Immunomodulatory drugs inhibit TLR4-induced type-1 interferon production independently of Cereblon via suppression of the TRIF/IRF3 pathway.
Immunomodulatory drugs inhibit TLR4-induced type-1 interferon production independently of Cereblon via suppression of the TRIF/IRF3 pathway.
International immunology (2016-02-13)
David Millrine, Haruhiko Miyata, Mami Tei, Praveen Dubey, Kishan Nyati, Taisuke Nakahama, Yohannes Gemechu, Barry Ripley, Tadamitsu Kishimoto
ABSTRACT
Thalidomide and its derivatives, collectively referred to as immunomodulatory drugs (IMiDs), are effective inhibitors of inflammation and are known to inhibit TLR-induced TNFα production. The identification of Cereblon as the receptor for these compounds has led to a rapid advancement in our understanding of IMiD properties; however, there remain no studies addressing the role of Cereblon in mediating the suppressive effect of IMiDs on TLR responses. Here, we developed Cereblon-deficient mice using the CRISPR-Cas9 system. TLR-induced cytokine responses were unaffected by Cereblon deficiency in vivo Moreover, IMiD treatment inhibited cytokine production even in the absence of Cereblon. The IMiD-induced suppression of cytokine production therefore occurs independently of Cereblon in mice. Further investigation revealed that IMiDs are potent inhibitors of TLR-induced type-1 interferon production via suppression of the TRIF/IRF3 pathway. These data suggest that IMiDs may prove effective in the treatment of disorders characterized by the ectopic production of type-1 interferon. Significantly, these properties are mediated separately from thalidomide's teratogenic receptor, Cereblon. Thus, certain therapeutic properties of Thalidomide can be separated from its harmful side effects.
Sigma-Aldrich
Pomalidomide, ≥98% (HPLC)
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by phenol extraction