Merck
  • Infection and characterization of Toxoplasma gondii in human induced neurons from patients with brain disorders and healthy controls.
Infection and characterization of Toxoplasma gondii in human induced neurons from patients with brain disorders and healthy controls.
Microbes and infection (2015-10-04)
Eleonora Passeri, Lorraine Jones-Brando, Claudia Bordón, Srona Sengupta, Ashley M Wilson, Amedeo Primerano, Judith L Rapoport, Koko Ishizuka, Shin-ichi Kano, Robert H Yolken, Akira Sawa
ABSTRACT
Toxoplasma gondii is a protozoan parasite capable of establishing persistent infection within the brain. Serological studies in humans have linked exposure to Toxoplasma to neuropsychiatric disorders. However, serological studies have not elucidated the related molecular mechanisms within neuronal cells. To address this question, we used human induced neuronal cells derived from peripheral fibroblasts of healthy individuals and patients with genetically-defined brain disorders (i.e. childhood-onset schizophrenia with disease-associated copy number variations). Parasite infection was characterized by differential detection of tachyzoites and tissue cysts in induced neuronal cells. This approach may aid study of molecular mechanisms underlying individual predisposition to Toxoplasma infection linked to neuropathology of brain disorders.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Nunc® Lab-Tek® Chamber Slide system, 4 wells, Permanox® slide, 1.8 cm2/well, sterile, 96/cs
Sigma-Aldrich
ECM Gel from Engelbreth-Holm-Swarm murine sarcoma, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Valproic acid sodium salt, 98%
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥96% (HPLC), powder